Membrane filters Nuclepore™ track etched, Whatman™
Supplier: Whatman products (Cytiva)
10417012
WHAT10417012EA 802 SGD
WHAT10417012
Membrane filters Nuclepore™ track etched, Whatman™
Filters Membrane Filters
Nuclepore™ track-etched hydrophilic membrane filters offer a highly defined pore size cut-off in the submicron range, enabling particle deformity measurement and capture of extremely small particles. The membranes have sharply defined pores, high flow rates, and excellent chemical and thermal resistance. They are suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
  • Track-etched for highly defined pore size and high pore density
  • Treated with polyvinylpyrrolidone (PVP) to
More Product Information