Quantitative filter papers, ashless grades, grade 43, Whatman™
Supplier: Whatman products (Cytiva)
1443-090
WHAT1443-090EA 101 SGD
WHAT1443-090
Quantitative filter papers, ashless grades, grade 43, Whatman™
Filters Filter Papers Quantitative Filter Paper
Cytiva's Whatman Grade 43 filters support inorganic analysis and other forms of instrumental analyses. The low ash content also makes Grade 43 appropriate for gravimetric analyses requiring sample combustion.
  • Very low ash content (0,007% ash concentration maximum)
  • Intermediate particle retention rating of 16 µm
  • Herzberg filtration speed of 155 s
  • Particle collection between that of Grades 40 and 41
Cytiva offers other ashless grades to ensure that you have the appropriate particle retention rating for your needs.
Particle collection and inorganicMore Product Information