Syringe filters, PES, Puradisc™, Whatman™
Supplier: Whatman products (Cytiva)
6781-2502
WHAT6781-2502EA 2960 SGD
WHAT6781-2502
Syringe filters, PES, Puradisc™, Whatman™
Filters Syringe Filters
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample typesMore Product Information