WHEATON® Boston Round Bottles, Clear, Narrow Mouth
Supplier: DWK Life Sciences
14235-666
WHEA14235-666EA 462 SGD
WHEA14235-666
WHEATON® Boston Round Bottles, Clear, Narrow Mouth
Bottles Glass Bottles
Wheaton Boston round bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.
These narrow mouth bottles facilitate pouring and storing of liquids. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. Partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Available with caps attached or in bulkMore Product Information