Ampoules, pre-scored, WHEATON®
Supplier: DWK Life Sciences
176792 176796 176772 176782
WHEA176792EA 474 SGD
WHEA176792 WHEA176796 WHEA176772 WHEA176782
Ampoules, pre-scored, WHEATON®
Ampoules
Glass ampoules made from low extractable borosilicate glass can be hermetically sealed to preserve purity and extend content shelf life.
  • Tamper-evident seal
  • For biological product preservation
  • Gold band on the stem of the ampoule indicates that it is pre-scored to facilitate snapping off the top eliminating the need to file the ampoule
Ideal for standards packaging, environmental and diagnostic standards, lyophilisation and cryogenic storage.
Certifications: USP Type I, ASTM E438 Type I, Class A
Order Now
SPECIFICATIONS
Environmentally Preferable