Arsine generator apparatus, Clear-Seal™, WHEATON®
Supplier: DWK Life Sciences
180020
WHEA180020EA 447 SGD
WHEA180020
Arsine generator apparatus, Clear-Seal™, WHEATON®
Digestion Systems Trace Element Detection Apparatus
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method.
Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Certifications: Meets ASTM / USP and EPA specifications.