Arsine generator apparatus, WHEATON®
Supplier: DWK Life Sciences
180030
WHEA180030EA 945 SGD
WHEA180030
Arsine generator apparatus, WHEATON®
Digestion Systems Trace Element Detection Apparatus
Designed for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method.
  • Assembled apparatus is freestanding and easy to use
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.