Reagent bottles, with screw cap, WHEATON®
Supplier: DWK Life Sciences
220283
WHEA220283EA 337 SGD
WHEA220283
Reagent bottles, with screw cap, WHEATON®
Bottles Glass Bottles
Manufactured from USP Type I borosilicate glass, with large writing patch.
  • Features no-drip pour lip
  • Pre-attached polypropylene screw cap with PTFE liner
  • Autoclavable