Bottles, narrow neck, glass, safety coated, clear/amber, round, WHEATON®
Supplier: DWK Life Sciences
220946 220955
WHEA220946EA 477 SGD
WHEA220946 WHEA220955
Bottles, narrow neck, glass, safety coated, clear/amber, round, WHEATON®
Bottles Glass Bottles
Bottles, safety coated with PVC-based plastisol.Made from clear soda-lime glass that conforms to USP Type III requirements.
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Autoclaving / sterilizing bottle is not recommended
  • Very good chemical resistance
  • Amber coloured for light sensitive substances
Plastisol coats exterior of bottle retaining contents if bottle is accidentally broken.
Certifications: These bottles help meet OSHA requirements
Order Now
SPECIFICATIONS
Environmentally Preferable