WHEATON® Serum Vials
Supplier: DWK Life Sciences
223685 223687
WHEA223685EA 283 SGD
WHEA223685 WHEA223687
WHEATON® Serum Vials
Vials
These clear glass vials are manufactured from Wheaton '200' low extractable borosilicate glass to maintain uniform wall thickness. The amber vials are manufactured from Wheaton '320'  borosilicate glass.
  • Blowback feature in the neck and a specially designed bottom radius for added strength
  • These features are important for freeze-drying applications
  • Vials are provided in shrink-wrapped modules which greatly reduces particulate contamination
Certifications: Meets ASTM Type 1, Class A/B and USP Type 1 standards.
Delivery information: VialsMore Product Information