Bottles, serum/sample, WHEATON®
Supplier: DWK Life Sciences
223742 223743 223745 223739 223748 223764 223766
WHEA223742EA 1040 SGD
WHEA223742 WHEA223743 WHEA223745 WHEA223739 WHEA223748 WHEA223764 WHEA223766
Bottles, serum/sample, WHEATON®
Vials
Serum bottles are ideal for long- and short-term sample storage, lyophilisation and vaccine/injectable drug containers. These moulded bottles have excellent wall strength and chemical tolerances which are important for storage applications. Bottles are manufactured from Wheaton 400 borosilicate moulded glass that conforms to USP Type I requirements. Bottles are packed in shrink-wrapped partitioned modules with the exception of 2 ml size, which has no partition.
  • Fits most freeze drying applications
  • Borosilicate glass conforms to USP Type I requirements
More Product Information