WHEATON® Aluminium Seals, Unlined
Supplier: DWK Life Sciences
224192-05 224193-01 224193-05 224193-06 224193-07 224183-01 224182-01 224178-01 224191_U 224177-01 224192-01 224191
WHEA224192-05EA 280 SGD
WHEA224192-05 WHEA224193-01 WHEA224193-05 WHEA224193-06 WHEA224193-07 WHEA224183-01 WHEA224182-01 WHEA224178-01 WHEA224191_U WHEA224177-01 WHEA224192-01 WHEA224191
WHEATON® Aluminium Seals, Unlined
Closures Chromatography Vial Closures
Aluminum seals (unlined) are available in a wide range of styles and sizes.
  • Open top seals expose stopper for sample retrieval with a syringe
  • Tear-off seal removes completely allowing content to pour from bottle
  • Solid top seals are excellent for long-term storage of samples
  • Flip cap seals provide tamper evidence
Colour selection includes natural, red, blue, and green.