Crimpers and decappers, WHEATON®
Supplier: DWK Life Sciences
224323
WHEA224323EA 1660 SGD
WHEA224323
Crimpers and decappers, WHEATON®
Crimpers, Cappers and Decappers
These crimpers and decappers are used to attach and detach 30 mm aluminium seals.