Sample vials in Lab File®, WHEATON®
Supplier: DWK Life Sciences
224802 224804 224831 224812 W224815
WHEA224802EA 336 SGD
WHEA224802 WHEA224804 WHEA224831 WHEA224812 WHEAW224815
Sample vials in Lab File®, WHEATON®
Vials
For storage of reagents and standards. Excellent for small derivatisation reactions. Wheaton 33® low extractable borosilicate glass is ideal for research studies where stability of glass is of prime importance.
Ordering information: Without cap.