WHEATON® Vials with Inserts
Supplier: DWK Life Sciences
225180 225185
WHEA225180EA 748 SGD
WHEA225180 WHEA225185
WHEATON® Vials with Inserts
Vials
These vials are made of durable PP and can be used in standard gas and liquid chromatography autosamplers.
  • Lightweight and economical
  • Available with snap/crimp or screw top finish
  • Conical interior ensures maximum retrieval of contents without the hassle of using removable inserts
Packaging: 100 vials per shelf pack or 1000 vials per case.
Order Now
SPECIFICATIONS
Environmentally Preferable