BOD bottles, WHEATON®
Supplier: DWK Life Sciences
227497-00G
WHEA227497-00GEA 1220 SGD
WHEA227497-00G
BOD bottles, WHEATON®
Bottles BOD Bottles
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
Order Now
SPECIFICATIONS
Environmentally Preferable