Tissue grinders, Dounce type
Supplier: DWK Life Sciences
357544
WHEA357544EA 1050 SGD
WHEA357544
Tissue grinders, Dounce type
Homogenisers Tissue Grinders
Borosilicate glass. Designed to retain a high percentage of cell nuclei and mitochondria in soft tissues. In operation, the pestle ball is encircled in liquid which avoids heat build up. Friction is reduced and there is reduced heat accumulation.
  • Mortar has large reservoir and pouring lip
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Delivery information: Each mortar is suppliedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable