Tissue grinders, Potter-Elvehjem type
Supplier: DWK Life Sciences
357966 357971 357976 357981 357986 357991
WHEA357966EA 454 SGD
WHEA357966 WHEA357971 WHEA357976 WHEA357981 WHEA357986 WHEA357991
Tissue grinders, Potter-Elvehjem type
Homogenisers Tissue Grinders
Mortar in borosilicate glass, PTFE pestle with stainless steel rod. Serrated PTFE pestle disperses homogenate into mortar cylinder more efficiently. Precisely constructed with completely interchangeable parts.
  • Clearance between pestle and tube is 0.1 to 0.15 mm
  • Stainless steel rod Ø 6.3 mm
  • Can be autoclaved
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).