Tissue grinders, Potter-Elvehjem type
Supplier: DWK Life Sciences
358034
WHEA358034EA 384 SGD
WHEA358034
Tissue grinders, Potter-Elvehjem type
Homogenisers Tissue Grinders
Mortar in borosilicate glass, PTFE pestle with stainless steel rod. Precisely constructed with completely interchangeable parts. For use in preparation of tissue homogenates, etc. PTFE pestle for soft tissue such as brain or liver.
  • Clearance between pestle and tube is 0,1 to 0,15 mm
  • Can be autoclaved
Stainless steel rod Ø 6,3 mm can be used with overhead stirrer (however, we strongly recommend using safety coated tissue grinders for added user safety).
Stainless steel rod Ø 6.3 mm can be used with overhead stirrer (however, we stronglyMore Product Information