Tissue grinders, conical
Supplier: DWK Life Sciences
358103 358109 358113 358105 358101
WHEA358103EA 447 SGD
WHEA358103 WHEA358109 WHEA358113 WHEA358105 WHEA358101
Tissue grinders, conical
Homogenisers Tissue Grinders
Borosilicate glass. Handheld homogeniser with conical surface, which allows initial size reduction, followed by passage through the cylindrical section for the final homogenisation step. Grinding efficiency is improved and less time is required as compared to the Tenbroeck and Potter-Elvehjem designs. Recommended for heart and lung tissues, as well for skin and plant tissues.
  • Clearance between pestle and tube is 0,1 to 0,15 mm
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: WheatonMore Product Information