WHEATON® Filtration Assembly with Stainless Steel Support and Standard Taper 40/35 Joint Connection, 47 mm
Supplier: DWK Life Sciences
419390
WHEA419390EA 1330 SGD
WHEA419390
WHEATON® Filtration Assembly with Stainless Steel Support and Standard Taper 40/35 Joint Connection, 47 mm
Funnels Filtration Funnels Filtration Apparatus Glass
This filtration assembly are designed for routine filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents.
  • Glass components are manufactured from WHEATON 33 low extractable borosilicate glass
  • Ground glass connection eliminates phthalate contamination that can occur from silicone or neoprene stoppers
  • Drip tip on the support bases extends below the hose connection preventing filtrate from entering the vacuum line
  • Recommended for filtering viscous or proteinaceous solutions or to produce ultraclean filtrate
Order Now
SPECIFICATIONS
Environmentally Preferable