Bottle top dispenser, Calibrex™ 520, WHEATON®
Supplier: DWK Life Sciences
844000
WHEA844000EA 4310 SGD
WHEA844000
Bottle top dispenser, Calibrex™ 520, WHEATON®
Bottle Top Dispensers
Variable volume.
  • Instant volume setting and chemical resistance
  • Long-lasting performance stability
  • Simple construction: No tool for disassembling
Designed for performance handling of liquids from a large variety of bottles and flasks, it combines the latest in dosing technology, high-tech materials and ergonomic design. Manufactured and tested to fully comply with current safety regulations.