Vial racks, WHEATON®
Supplier: DWK Life Sciences
868805
WHEA868805EA 468 SGD
WHEA868805
Vial racks, WHEATON®
Racks Chromatography Vial Racks
These polypropylene racks accommodate 36 or 50 headspace auto-sampler vials. Each well has an opening in the bottom to facilitate drainage.
  • Autoclavable and stackable
  • Chemically resistant
  • Include position identifiers