Staining dishes, WHEATON®
Supplier: DWK Life Sciences
900202
WHEA900202EA 213 SGD
WHEA900202
Staining dishes, WHEATON®
Jars Staining Jars
Soda-lime glass and stainless steel.
  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.