Staining jars according to Coplin, WHEATON®
Supplier: DWK Life Sciences
900570
WHEA900570EA 83.2 SGD
WHEA900570
Staining jars according to Coplin, WHEATON®
Jars Staining Jars
Soda-lime glass.
  • Linerless, white PP screw cap to reduce solvent evaporation
  • Capacity: Approx. 60 ml
This unit is used for staining slides, or as a developing chamber for thin layer chromatography. It holds five single 76×26 mm slides vertically, or 10 slides back to back.
Order Now
SPECIFICATIONS
Environmentally Preferable