WHEATON® Scintillation Vials
Supplier: DWK Life Sciences
986546 986568 986542 986541
WHEA986546EA 1020 SGD
WHEA986546 WHEA986568 WHEA986542 WHEA986541
WHEATON® Scintillation Vials
Vials
These vials are made from WHEATON® 180 low potassium borosilicate glass. The background counts of these vials are consistently low, the ultraviolet transmission is high.
  • Available with attached or unattached screw caps
  • Utility trays can be used to store samples
Certifications: Glass conforms to ASTM E438 Type I, Class A.
Packaging: Packed in five utility trays with each partition tray holding 100 vials.