Scintillation vials, PET, WHEATON®
Supplier: DWK Life Sciences
986731
WHEA986731EA 664 SGD
WHEA986731
Scintillation vials, PET, WHEATON®
Vials
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
  • Clarity of glass
  • Can be safely incinerated
  • Lightweight and virtually unbreakable
* With cap on