Bottles, narrow neck, round, Boston type, WHEATON®
Supplier: DWK Life Sciences
W216845 W216846 W216842 W216843 W216844 W216814
WHEAW216845EA 73.8 SGD
WHEAW216845 WHEAW216846 WHEAW216842 WHEAW216843 WHEAW216844 WHEAW216814
Bottles, narrow neck, round, Boston type, WHEATON®
Bottles Glass Bottles
Soda lime glass, Class III, clear or amber.
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Polypropylene or phenolic caps with choice of liners
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Ideal for solvent, chemical or sample storage. Amber coloured for light-sensitive materials.
Order Now
SPECIFICATIONS
Environmentally Preferable