WHEATON® Jars, Wide Mouth, with Thread
Supplier: DWK Life Sciences
W216904 W216920 W216921
WHEAW216904EA 129 SGD
WHEAW216904 WHEAW216920 WHEAW216921
WHEATON® Jars, Wide Mouth, with Thread
Jars General Use Jars
Jars, made of clear soda-lime glass, straight sided.
  • Wide mouth design is great for large solid samples
  • Walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • Available with caps attached or bulk packs without caps
Certifications: Conforms to USP Type III requirements.