Bottles, wide neck, with screw cap, WHEATON®
Supplier: DWK Life Sciences
W216929
WHEAW216929EA 163 SGD
WHEAW216929
Bottles, wide neck, with screw cap, WHEATON®
Bottles Glass Bottles
Made of type III soda-lime glass, clear.
  • Largest bottles available
  • Very good chemical resistance
  • PP screw cap with poly-vinyl liner or PTFE lined screw cap
Ideal for general, small to large volume storage. Hazardous communication label included with each case of bottles allows for content ID and safety notification. Tamper-evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse.