Bottles, wide neck, graduated, Valumetric®, WHEATON®
Supplier: DWK Life Sciences
W216970
WHEAW216970EA 211 SGD
WHEAW216970
Bottles, wide neck, graduated, Valumetric®, WHEATON®
Bottles Glass Bottles
Soda-lime glass, class III, clear.
  • Ideal for general storage
  • Easy removal of contents
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper-evident seal included with each case of bottles keeps bottle sealed which protects contents from accidental misuse