WHEATON® Valumetric™ Graduated Bottle
Supplier: DWK Life Sciences
W216976
WHEAW216976EA 231 SGD
WHEAW216976
WHEATON® Valumetric™ Graduated Bottle
Bottles Glass Bottles
WHEATON® Graduated Valumetric™ bottles with black phenolic cap with poly-seal liner for storage and samples of solids, dry powdered materials and liquids.
  • Ideal for use as sample bottles
  • Useful for dry powders, soil samples, and specimen storage
  • Made from Wheaton 800 glass, which meets USP Type III standards
  • Graduated in ml and oz.
Autoclavable black phenolic cap with rubber liner.
Order Now
SPECIFICATIONS
Environmentally Preferable