WHEATON® Premium Rubber Stoppers
Supplier: DWK Life Sciences
W224100-406 W224100-408 W224100-402
WHEAW224100-406EA 1080 SGD
WHEAW224100-406 WHEAW224100-408 WHEAW224100-402
WHEATON® Premium Rubber Stoppers
Stoppers Rubber Stoppers
Ultra-pure rubber stoppers made from a bromobutyl rubber compound containing the highest level of chemical purity. These stoppers demonstrate very low levels of moisture absorption and extractables.
  • Compatible with Water For Injection (WFI) applications
  • Free from natural rubber or natural rubber latex
They are ideal for use in most lyophilisation applications. Complete coat rubber stoppers are completely coated with a fluorinated polymer that has barrier properties similar to PTFE.
The fluorinated polymer coating is all that comes in contact withMore Product Information