WHEATON® Sample Vials in Lab File®
Supplier: DWK Life Sciences
W224581 W224584 W224589 W224590 W224591 W224681 W224684 W224840
WHEAW224581EA 985 SGD
WHEAW224581 WHEAW224584 WHEAW224589 WHEAW224590 WHEAW224591 WHEAW224681 WHEAW224684 WHEAW224840
WHEATON® Sample Vials in Lab File®
Vials
Clear vials made from low extractable borosilicate glass, amber vials made from borosilicate glass.
  • Sample vials with caps attached to vial help maintain cleanliness
  • Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • Clear vials conform to ASTM E438 Type I, Class A and USP Type I requirements
  • Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products
  • 14B rubber lined black phenolic screw cap for aqueous samples
Order Now
SPECIFICATIONS
Environmentally Preferable