High performance crimping tool and jaw sets, WHEATON®
Supplier: DWK Life Sciences
W225762 W225763
WHEAW225762EA 3110 SGD
WHEAW225762 WHEAW225763
High performance crimping tool and jaw sets, WHEATON®
Crimpers, Cappers and Decappers
These high performance crimping tools are designed with inter-changeable jaws, so that users only need one tool, and can purchase individually sized crimpers and decappers for specific laboratory requirements.
  • Designed with external power source and cord (no battery)
  • Optional base and mounting kit
The jaw sets are available as crimper and decappers, and fit standard lab vials seals with aluminum or two-part aluminum/steel caps. The crimping tool has the ability to store upto nine programs for different caps and cap sizes. The tools are ergonomicallyMore Product Information