Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
Supplier: DWK Life Sciences
W240846 W240836 W240850 W240837
WHEAW240846EA 639 SGD
WHEAW240846 WHEAW240836 WHEAW240850 WHEAW240837
Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
Closures Test/Sample Tube Closures
Choice of black phenolic, white polypropylene or urea screw caps
  • PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product
  • Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals
Order Now
SPECIFICATIONS
Environmentally Preferable