Deck kit for small bottle roller culture system, Wheaton®
Supplier: DWK Life Sciences
W348930-CH
WHEAW348930-CHEA 1710 SGD
WHEAW348930-CH
Deck kit for small bottle roller culture system, Wheaton®
Roller Culture Apparatus
Attaches quickly and easily to the small bottle bench top roller culture system. Holds an additional two rollers per kit. These units will accept two additional decks. Kit consists of assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions.
Weight: 2,3 kg