Lyophilisation ampoules, pre-scored, Vacule®, WHEATON®
Supplier: DWK Life Sciences
W651506
WHEAW651506EA 1520 SGD
WHEAW651506
Lyophilisation ampoules, pre-scored, Vacule®, WHEATON®
Ampoules
Made from low extractable borosilicate glass
  • Pre-scored for easy opening
  • Heat sealed only
These ampoules are ideal for small volume lyophilisation samples.
Certifications: USP Type I, ASTM E 438 Type I, Class A
Order Now
SPECIFICATIONS
Environmentally Preferable