Cryogenic freezer boxes, CryoFile™, WHEATON®
Supplier: DWK Life Sciences
W651603 W651603-XL
WHEAW651603EA 339 SGD
WHEAW651603 WHEAW651603-XL
Cryogenic freezer boxes, CryoFile™, WHEATON®
Boxes Cryoboxes
High quality chip-board, spray coated with polyethylene.
  • Water repellent material allows for longer durability
  • Numbering system printed on lid and bottom of the box
  • Withstands cryogenic temperatures to –196 °C
  • Six colours provide easy sample idenfication
Use for batching and storing cryogenic vials or ampoules. Partitions numbered from 1 to 81 for easy content identification.
Order Now
SPECIFICATIONS
Environmentally Preferable