Cryogenic vials, CryoELITE®, WHEATON®
Supplier: DWK Life Sciences
W985868 W985865 W985863 W985872
WHEAW985868EA 697 SGD
WHEAW985868 WHEAW985865 WHEAW985863 WHEAW985872
Cryogenic vials, CryoELITE®, WHEATON®
Vials
CryoELITE® cryogenic vials are made from low binding, cryogenic grade virgin PP. They offer an unrivaled cap seal for the shipping and storage of precious biosamples. The patented cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling.
  • Loctagon™ Vial Skirt provides stability in freestanding position
  • Screw cap can be easily removed with one hand
  • Coloured caps allow for colour coding
  • Round bottom vials can be centrifuged up to 17 000×g
Optional 2D data matrix barMore Product Information