Bioreactors, Wheaton® CELLine™
Supplier: DWK Life Sciences
WCL1000-3
WHEAWCL1000-3EA 2690 SGD
WHEAWCL1000-3
Bioreactors, Wheaton® CELLine™
Bioreactors
CELLine™ is a disposable, two-compartment bioreactor manufactured from optically clear virgin PS with a gas transfer bottom made of a moulded silicone membrane providing a 0,2 µm vent barrier. The compartments are separated by a 10 kDa semi-permeable cellulose acetate membrane and individually pressure tested for integrity. CELLine™ classic (CL) is ideal for laboratory scale applications using suspension cells or adherent cells in combination with microcarriers. The unit is optimised for cultivation of hybridomas and many other cell types (for example CHO, NSO, SF cells). CELLine™ adhere (AD)More Product Information