Gas cylinder change-over unit, MK3 series
Supplier: WEST GROUP
PC30
WSTGPC30EA 5840 SGD
WSTGPC30
Gas cylinder change-over unit, MK3 series
Gas Regulators
Pneuchange is the unit of choice in thousands of laboratories for ensuring the continuity of C0₂ and other inert gases supplies to C0₂ incubators and other equipment where loss of gas would be inconvenient, costly or dangerous.
  • Suitable for use with all non-corrosive and non flammable gases
  • Bench or wall mounted using the keyholes on the back of the unit
  • An audible battery operated change-over alarm is fitted
The inlet ports are connected with suitable tubing to the gas bottle regulators. Two pressure gauges indicate the inlet pressures and twoMore Product Information