Thermoreactors, CR series
Supplier: WTW
1P21-1
WTWA1P21-1EA 4380 SGD
WTWA1P21-1
Thermoreactors, CR series
Block Heaters Digital Block Heaters
Thermoreactors for the determination of COD, total nitrogen or total phosphorus, ensure complete digestion of the sample and maintain the necessary high reaction temperature throughout the defined period. The most important temperatures and digestion times are stored in 8 fixed, easily selectable digestion programmes. CR 3200 and CR 4200 can be used with optional TFK CR external temperature probe, which can be used as a testing aid. It can be plugged into the interface in place of a cuvette, the set and actual temperatures can be output to either a printer or a PC, so function can not onlyMore Product Information