Masterflex® L/S® High-Performance Precision Pump Tubing, FDA-Compliant Viton®, L/S 35; 25 ft
Supplier: Avantor Fluid Handling
VWR Item#MFLA96412-35
Your Organization's Price:
Retrieving
Good for food and lab applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 35
Applications: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work
Characteristics: Good chemical resistance, opaque black