Masterflex® L/S® Precision Pump Tubing, Tygon® E-Food, L/S 16; 50 ft
Supplier: Avantor Fluid Handling
VWR Item#MFLA06418-16
Your Organization's Price:
Retrieving
Designed specifically for handling food products.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 16
Applications: Food and beverage processing or dispensing
Characteristics: Nonwetting, odorless, tasteless, nontoxic, transparent