Gauge Guard Isolator Seal, EPDM; 1-1/2" Tri-Clamp
Supplier: Avantor Fluid Handling
VWR Item#MFLA30565-19
Your Organization's Price:
Retrieving
Extend the life and accuracy of your expensive and fragile instruments.
  • Manufactured to meet USP Class VI certification
  • Animal derived ingredient free
<B>Gauge Guard Protectors</B> safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals. <br /><br /><B>Gauge Guard Isolator Seals</B> combine a hygenic seal with a membrane to isolate hot or caustic solutions from your instrument. Available in platinum-cured silicone or EPDM.