Masterflex® Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/Pk
Supplier: Avantor Fluid Handling
VWR Item#MFLA06833-01
Your Organization's Price:
Retrieving
Won't rust, corrode, or distort.
  • Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.