Masterflex® Flow-Through Tubing Weight; Stainless Steel
Supplier: Avantor Fluid Handling
VWR Item#MFLA07570-04
Your Organization's Price:
Retrieving
Keep your samples flowing and tubing from curling.
  • Choose between PTFE or stainless steel construction
  • Securely anchor tubing in receiving vessel during dispensing or fluid transfer applications
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.
Flow-through PTFE tubing weight anchors tubing in receiving vessel. Accepts tubing with ID of 5/16'' to 7/16''. Fits Masterflex pump tubing sizes L/S® 17, L/S 18, L/S 24, L/S 35, and L/S 36.