Masterflex® Panel Mount Flow Controller, Pulse Input, 4-20 mA Output
Supplier: Avantor Fluid Handling
VWR Item#MFLA33110-60
Your Organization's Price:
Retrieving
Easily control, view, and store your flow rates.
  • Control your Low-Flow Impeller Sensors with these controllers
These new controllers display flow rate and total (resettable) simultaneously on two 8-digit Display LCDs. Push-button keys and menu prompts make it easy to program the controller. The 4-to-20 mA output lets you send data to a recorder or datalogger. Nonvolatile memory stores flow total in the event of power failure. <br /><br />Select a 1¼4 DIN panel- or wall-mount controller depending on your application needs. Panel-mount model 33110-60 features a gasketed front panel, membrane switches, and NEMA Type 12 (IP52) enclosure. Wall-mount model 33110-70 has a clear cover and splashproof NEMA Type 4X (IP56) enclosure. Both models include a 21¼2-ft L cable with stripped ends. Order power supply for 110 VAC operation separately.