Masterflex® Fitting, Polypropylene, Tee, Female Luer to Female Luer to Female Luer Adapter; 25/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLA45508-56
Your Organization's Price:
Retrieving
Full material traceability to ensure process integrity.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100000 environment
  • Connection Type: Female Luer to Female Luer to Female Luer
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.